Chemical Identifiers
CAS53213-94-8
IUPAC Name3-propyl-2-[(3E)-5-[(2Z)-3-propyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular FormulaC25H27IN2S2
InChI KeyGDEURKKLNUGTDA-UHFFFAOYSA-M
SMILES[I-].CCCN1C(SC2=CC=CC=C12)=CC=CC=CC1=[N+](CCC)C2=CC=CC=C2S1
View more