Thermo Scientific Chemicals

3-(Di-tert-butylphosphonium)propane sulfonate, 97%, Thermo Scientific Chemicals

Catalog number: H34253.03
1 g, Each
Thermo Scientific Chemicals

3-(Di-tert-butylphosphonium)propane sulfonate, 97%, Thermo Scientific Chemicals

Catalog number: H34253.03
1 g, Each
Quantity
Catalog number: H34253.03
also known as H34253-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1055888-89-5
IUPAC Name
3-(di-tert-butylphosphanyl)propane-1-sulfonic acid
Molecular Formula
C11H25O3PS
InChI Key
JPNPRWMRUCIEMN-UHFFFAOYSA-N
SMILES
CC(C)(C)P(CCCS(O)(=O)=O)C(C)(C)C
Appearance (Color)
White
Form
Powder
Assay by NMR
>96.0%
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure

Description

3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text