Thermo Scientific Chemicals

3-Iodochromone, 97%, Thermo Scientific Chemicals

Catalog number: H34295.03
1 g, Each
Thermo Scientific Chemicals

3-Iodochromone, 97%, Thermo Scientific Chemicals

Catalog number: H34295.03
1 g, Each
Quantity
Catalog number: H34295.03
also known as H34295-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122775-34-2
IUPAC Name
3-iodo-4H-chromen-4-one
Molecular Formula
C9H5IO2
InChI Key
SZDBDDKQJHHRGH-UHFFFAOYSA-N
SMILES
IC1=COC2=CC=CC=C2C1=O
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
92.0-101.0?C
Appearance (Color)
Pale yellow

Description

3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agent, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text