Thermo Scientific Chemicals

1-Bromomethyl-2,2-difluorocyclopropane, 97%, Thermo Scientific Chemicals

Catalog number: H34305.MD
250 mg, Each
Thermo Scientific Chemicals

1-Bromomethyl-2,2-difluorocyclopropane, 97%, Thermo Scientific Chemicals

Catalog number: H34305.MD
250 mg, Each
Quantity
Catalog number: H34305.MD
also known as H34305-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77613-65-1
IUPAC Name
2-(bromomethyl)-1,1-difluorocyclopropane
Molecular Formula
C4H5BrF2
InChI Key
TUDJNSKRXIUOAJ-UHFFFAOYNA-N
SMILES
FC1(F)CC1CBr
Identification (FTIR)
Conforms
Refractive Index
1.4220-1.4270 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text