Thermo Scientific Chemicals

2-Ethoxybenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: H34325.06
5 g, Each
Thermo Scientific Chemicals

2-Ethoxybenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: H34325.06
5 g, Each
Quantity
Catalog number: H34325.06
also known as H34325-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68800-33-9
IUPAC Name
2-ethoxybenzene-1-sulfonyl chloride
Molecular Formula
C8H9ClO3S
InChI Key
RWURYJNJYODVAL-UHFFFAOYSA-N
SMILES
CCOC1=CC=CC=C1S(Cl)(=O)=O
Appearance (Color)
White to pale pink
Form
Crystals or powder or crystalline powder
Assay (GC)
≥95.0%
Melting Point (clear melt)
59.0-66.0?C

Description

2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text