2,6-Difluoro-4-methoxypyridine, 97%, Thermo Scientific Chemicals
2,6-Difluoro-4-methoxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Difluoro-4-methoxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H34330.03
also known as H34330-03
Price (USD)/ Each
162.65
Online exclusive
180.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
162.65
Online exclusive
180.00 
Save 17.35 (10%)
Add to cart
2,6-Difluoro-4-methoxypyridine, 97%, Thermo Scientific Chemicals
Catalog numberH34330.03
Price (USD)/ Each
162.65
Online exclusive
180.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1184172-35-7
IUPAC Name2,6-difluoro-4-methoxypyridine
Molecular FormulaC6H5F2NO
InChI KeyHRTFKBJLWDISRQ-UHFFFAOYSA-N
SMILESCOC1=CC(F)=NC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only