1H-Indene-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1H-Indene-3-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1H-Indene-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34344.03
also known as H34344-03
Price (USD)/ Each
119.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
119.00
Add to cart
1H-Indene-3-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH34344.03
Price (USD)/ Each
119.00
-
Add to cart
Chemical Identifiers
CAS5020-21-3
IUPAC Name1H-indene-1-carboxylic acid
Molecular FormulaC10H8O2
InChI KeyKGWYICAEPBCRBL-UHFFFAOYNA-N
SMILESOC(=O)C1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)153.0-162.0?C
Appearance (Color)Yellow
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only