Thermo Scientific Chemicals

7-Bromo-5-methylbenzo[b]furan, 97%, Thermo Scientific Chemicals

Catalog number: H34347.MD
250 mg, Each
Thermo Scientific Chemicals

7-Bromo-5-methylbenzo[b]furan, 97%, Thermo Scientific Chemicals

Catalog number: H34347.MD
250 mg, Each
Quantity
Catalog number: H34347.MD
also known as H34347-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35700-48-2
IUPAC Name
7-bromo-5-methyl-1-benzofuran
Molecular Formula
C9H7BrO
InChI Key
TZUMDTURHSPEAY-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=C2OC=CC2=C1
Refractive Index
1.5935-1.5985 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text