Invitrogen™

HCS LipidTOX™ Green Phospholipidosis Detection Reagent (1000X), for cellular imaging

Catalog number: H34350
Have Questions?
Invitrogen™

HCS LipidTOX™ Green Phospholipidosis Detection Reagent (1000X), for cellular imaging

Catalog number: H34350
Have Questions?
Catalog Number
H34350
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
H343501 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The HCS LipidTOX™ Green phospholipidosis detection reagent was developed to characterize the potentially toxic side effects of compounds on lipid metabolism in mammalian cell lines. This reagent is easily detected by fluorescence microscopy or an HCS reader equipped with filters that can detect excitation/emission maxima ∼495/525 nm. LipidTOX™ phospholipidosis detection reagents do not affect the normal growth of cells, and the staining observed after incubation in live cells is maintained after formaldehyde fixation. This probe is compatible with the HCS LipidTOX™ neutral lipid stains (H34475, H34476, and H34477).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Equipment)
High Content Analysis Instrument
Product Type
Phospholipidosis Detection Reagent
Dye Type
Other Label(s) or Dye(s)
Format
96-well plate
Product Line
LipidTOX™
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text