HCS LipidTOX™ Red Phospholipidosis Detection Reagent (1000X), for cellular imaging
HCS LipidTOX™ Red Phospholipidosis Detection Reagent (1000X), for cellular imaging
Invitrogen™

HCS LipidTOX™ Red Phospholipidosis Detection Reagent (1000X), for cellular imaging

The HCS LipidTOX™ Red phospholipidosis detection reagent was developed to characterize the potentially toxic side effects of compounds on lipidRead more
Have Questions?
Catalog NumberQuantity
H343511 each
Catalog number H34351
Price (EUR)
508,00
Each
-
Add to cart
Quantity:
1 each
Price (EUR)
508,00
Each
Add to cart
Ask our AI about this Product
The HCS LipidTOX™ Red phospholipidosis detection reagent was developed to characterize the potentially toxic side effects of compounds on lipid metabolism in mammalian cell lines. This reagent is easily detected by fluorescence microscopy or an HCS reader equipped with filters that can detect excitation/emission maxima ∼595/615 nm. LipidTOX™ phospholipidosis detection reagents do not affect the normal growth of cells, and the staining observed after incubation in live cells is maintained after formaldehyde fixation. This probe is compatible with the HCS LipidTOX™ neutral lipid stains (H34475, H34476, and H34477).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeOther Label(s) or Dye(s)
Format96-well plate
Quantity1 each
Shipping ConditionRoom Temperature
For Use With (Equipment)High Content Analysis Instrument
Product LineLipidTOX
Product TypePhospholipidosis Detection Reagent
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3158233Certificate of AnalysisApr 01, 2025H34351
3143074Certificate of AnalysisApr 01, 2025H34351
3079905Certificate of AnalysisNov 07, 2024H34351
2925588Certificate of AnalysisJul 21, 2024H34351
2845272Certificate of AnalysisMar 27, 2024H34351
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link