Thermo Scientific Chemicals

3-(2-Methylphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H34375.09
10 g, Each
Thermo Scientific Chemicals

3-(2-Methylphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H34375.09
10 g, Each
Quantity
Catalog number: H34375.09
also known as H34375-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22084-89-5
IUPAC Name
3-(2-methylphenyl)propanoic acid
Molecular Formula
C10H12O2
InChI Key
JIRKNEAMPYVPTD-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1CCC(O)=O
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (clear melt)
98.0-107.0?C
Assay (Silylated GC)
≥95.0%

Description

3-(2-Methylphenyl)propionic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methylphenyl)propionic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text