Thermo Scientific Chemicals

1-Methyl-3-indolemethylamine, 96%, Thermo Scientific Chemicals

Catalog number: H34418.MD
250 mg, Each
Thermo Scientific Chemicals

1-Methyl-3-indolemethylamine, 96%, Thermo Scientific Chemicals

Catalog number: H34418.MD
250 mg, Each
Quantity
Catalog number: H34418.MD
also known as H34418-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19293-60-8
IUPAC Name
1-(1-methyl-1H-indol-3-yl)methanamine
Molecular Formula
C10H12N2
InChI Key
NOFZMDGMQKRLIV-UHFFFAOYSA-N
SMILES
CN1C=C(CN)C2=CC=CC=C12
Form
Liquid
Refractive Index
1.6130-1.6190 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text