Thermo Scientific Chemicals

2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide, Thermo Scientific Chemicals

Catalog number: H34443.14
25 g, Each
Thermo Scientific Chemicals

2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide, Thermo Scientific Chemicals

Catalog number: H34443.14
25 g, Each
Quantity
Catalog number: H34443.14
also known as H34443-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14077-58-8
IUPAC Name
2-ethoxyacetyl chloride
Molecular Formula
C4H7ClO2
InChI Key
ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
SMILES
CCOCC(Cl)=O
Assay (GC)
>96.0%
Appearance (Color)
Clear, colourless to pale yellow
Assay (Titration ex Chloride)
>96.0%
Refractive Index
1.4175-1.4225
Form
liquid

Description

It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

Solubility
Insoluble in water.

Notes
Reacts with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text