Thermo Scientific Chemicals

6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals

Catalog number: H34453.03
1 g, Each
Thermo Scientific Chemicals

6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals

Catalog number: H34453.03
1 g, Each
Quantity
Catalog number: H34453.03
also known as H34453-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6188-23-4
IUPAC Name
6-bromoimidazo[1,2-a]pyridine
Molecular Formula
C7H5BrN2
InChI Key
FXPMFQUOGYGTAM-UHFFFAOYSA-N
SMILES
BrC1=CN2C=CN=C2C=C1
Melting Point (clear melt)
76.0-82.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale yellow
Assay (GC)
≥97.5%

Description

It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

Solubility
slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text