Thermo Scientific Chemicals

2-Methoxybenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: H34455.03
1 g, Each
Thermo Scientific Chemicals

2-Methoxybenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: H34455.03
1 g, Each
Quantity
Catalog number: H34455.03
also known as H34455-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10130-87-7
IUPAC Name
2-methoxybenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO3S
InChI Key
GYOBZOBUOMDRRN-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1S(Cl)(=O)=O
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
50.5-59.5?C
Appearance (Color)
White to pale cream
Assay (GC)
≥94.0%

Description

For synthesis and pharmacological evaluation of sulfonamide derivatives of thiazolidin-4-ones 5-Bromo-2-methoxybenzenesulfonyl chloride is used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For synthesis and pharmacological evaluation of sulfonamide derivatives of thiazolidin-4-ones 5-Bromo-2-methoxybenzenesulfonyl chloride is used.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text