Thermo Scientific Chemicals

4'-Methylbiphenyl-4-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog number: H34459.03
1 g, Each
Thermo Scientific Chemicals

4'-Methylbiphenyl-4-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog number: H34459.03
1 g, Each
Quantity
Catalog number: H34459.03
also known as H34459-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
720-73-0
IUPAC Name
4'-methyl-[1,1'-biphenyl]-4-carboxylate
Molecular Formula
C14H11O2
InChI Key
RZOCCLOTCXINRG-UHFFFAOYSA-M
SMILES
CC1=CC=C(C=C1)C1=CC=C(C=C1)C([O-])=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (clear melt)
251.0-260.0?C
Form
Powder
Assay (Silylated GC)
≥95.0%

Description

The 4'-methyl-biphenyl-4-carboxylic acid exhibits the characteristic absorption bands of COOH and CH, groups. Photoproduct analysis, carried out by GC-MS, ETIR and GC-FTIR confirm three successive oxidation states of methyl suhstituents such as alcohol, aldehyde and carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The 4′-methyl-biphenyl-4-carboxylic acid exhibits the characteristic absorption bands of COOH and CH, groups. Photoproduct analysis, carried out by GC-MS, ETIR and GC-FTIR confirm three successive oxidation states of methyl suhstituents such as alcohol, aldehyde and carboxylic acids.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text