Thermo Scientific Chemicals

D-(+)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Catalog number: H35096.03
1 g, Each
Thermo Scientific Chemicals

D-(+)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Catalog number: H35096.03
1 g, Each
Quantity
Catalog number: H35096.03
also known as H35096-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7326-19-4
IUPAC Name
(2R)-2-hydroxy-3-phenylpropanoic acid
Molecular Formula
C9H10O3
InChI Key
VOXXWSYKYCBWHO-MRVPVSSYSA-N
SMILES
O[C@H](CC1=CC=CC=C1)C(O)=O
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Optical Rotation
+18.5? ± 1.5? (c=1 in Methanol)
Form
Crystals or powder or crystalline powder

Description

D-(+)-3-Phenyllactic acid is a chiral building block that is used in preparation of statine. Starting material in preparation of the hypoglycemic agent enlitazone and of (15)N-labeled phenylalanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-3-Phenyllactic acid is a chiral building block that is used in preparation of statine. Starting material in preparation of the hypoglycemic agent enlitazone and of (15)N-labeled phenylalanine.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text