Thermo Scientific Chemicals

4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione, Thermo Scientific Chemicals

Catalog number: H35120.ME
500 mg, Each
Thermo Scientific Chemicals

4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione, Thermo Scientific Chemicals

Catalog number: H35120.ME
500 mg, Each
Quantity
Catalog number: H35120.ME
also known as H35120-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64224-21-1
IUPAC Name
4-methyl-5-(pyrazin-2-yl)-3H-1,2-dithiole-3-thione
Molecular Formula
C8H6N2S3
InChI Key
CKNAQFVBEHDJQV-UHFFFAOYSA-N
SMILES
CC1=C(SSC1=S)C1=CN=CC=N1
Appearance (Color)
Red
Assay (HPLC)
> 97.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text