Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H35174.03
also known as H35174-03
Price (USD)/ Each
138.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
138.00
Add to cart
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH35174.03
Price (USD)/ Each
138.00
-
Add to cart
Chemical Identifiers
CAS859833-15-1
IUPAC Nameimidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Molecular FormulaC8H6Cl2N2O
InChI KeyLHLAJCRPSZDOFF-UHFFFAOYSA-N
SMILESCl.ClC(=O)C1=CN2C=CN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only