4-(3-Chloropropyl)morpholine, 95%, Thermo Scientific Chemicals
4-(3-Chloropropyl)morpholine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(3-Chloropropyl)morpholine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H35222.09
also known as H35222-09
Price (USD)/ Each
67.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
67.10
Add to cart
4-(3-Chloropropyl)morpholine, 95%, Thermo Scientific Chemicals
Catalog numberH35222.09
Price (USD)/ Each
67.10
-
Add to cart
Chemical Identifiers
CAS7357-67-7
IUPAC Name4-(3-chloropropyl)morpholine
Molecular FormulaC7H14ClNO
InChI KeyPIAZYBLGBSMNLX-UHFFFAOYSA-N
SMILESClCCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4705-1.4765 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only