Thermo Scientific Chemicals

3-Iodopyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H35255.03
1 g, Each
Thermo Scientific Chemicals

3-Iodopyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H35255.03
1 g, Each
Quantity
Catalog number: H35255.03
also known as H35255-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19363-99-6
IUPAC Name
3-iodopyrazolo[1,5-a]pyridine
Molecular Formula
C7H5IN2
InChI Key
PNNARKUCAKULAL-UHFFFAOYSA-N
SMILES
IC1=C2C=CC=CN2N=C1
Appearance (Color)
Brown
Assay (GC)
≥96.0%
Melting Point (clear melt)
63.5-72.5?C
Form
Powder

Description

Mostly used in the suzuki coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mostly used in the suzuki coupling reaction.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text