5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one, 98%, Thermo Scientific Chemicals
5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H35275.MD
also known as H35275-MD
Price (USD)/ Each
229.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
229.00
Add to cart
5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one, 98%, Thermo Scientific Chemicals
Catalog numberH35275.MD
Price (USD)/ Each
229.00
-
Add to cart
Chemical Identifiers
CAS198141-12-7
IUPAC Name5H,6H,7H,8H,9H-pyrido[3,2-b]indol-9-one
Molecular FormulaC11H10N2O
InChI KeyFATNUJIDKOPJRF-UHFFFAOYSA-N
SMILESO=C1CCCC2=C1C1=NC=CC=C1N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only