3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only
Jie Zhou; Jing Jin; Yi Zhang; Yuwen Yin; Xiaoguang Chen; Bailing Xu . Synthesis and antiproliferative evaluation of novel benzoimidazole-contained oxazole-bridged analogs of combretastatin A-4. European Journal of Medicinal Chemistry. 2013, 68, 222-232