Thermo Scientific Chemicals

(Iodomethyl)cyclopropane, 95%, Thermo Scientific Chemicals

Catalog number: H35309.03
1 g, Each
Thermo Scientific Chemicals

(Iodomethyl)cyclopropane, 95%, Thermo Scientific Chemicals

Catalog number: H35309.03
1 g, Each
Quantity
Catalog number: H35309.03
also known as H35309-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33574-02-6
IUPAC Name
(iodomethyl)cyclopropane
Molecular Formula
C4H7I
InChI Key
JHKJQWFHNIOUKY-UHFFFAOYSA-N
SMILES
ICC1CC1
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥94.0%
Refractive Index
1.5385-1.5445 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text