Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%, Thermo Scientific Chemicals

Catalog number: H35352.22
100 g, Each
Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%, Thermo Scientific Chemicals

Catalog number: H35352.22
100 g, Each
Quantity
Catalog number: H35352.22
also known as H35352-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
178631-04-4
IUPAC Name
3-methyl-1-octyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular Formula
C14H23F6N3O4S2
InChI Key
LECQXINNQGHJBM-UHFFFAOYSA-N
SMILES
CCCCCCCCN1C=C[N+](C)=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Appearance (Color)
Clear colorless to yellow to orange
Form
Viscous liquid
Identification (FTIR)
Conforms
Residual halide
<100ppm
Ion Chromatography
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text