Thermo Scientific Chemicals

Imidazo[1,2-a]pyridine-7-carbohydrazide, 95%, Thermo Scientific Chemicals

Catalog number: H35365.MD
250 mg, Each
Thermo Scientific Chemicals

Imidazo[1,2-a]pyridine-7-carbohydrazide, 95%, Thermo Scientific Chemicals

Catalog number: H35365.MD
250 mg, Each
Quantity
Catalog number: H35365.MD
also known as H35365-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
421595-78-0
IUPAC Name
imidazo[1,2-a]pyridine-7-carbohydrazide
Molecular Formula
C8H8N4O
InChI Key
VXJBLZXLLRYHPD-UHFFFAOYSA-N
SMILES
NNC(=O)C1=CC2=NC=CN2C=C1
Appearance (Color)
Cream
Form
Powder
Assay (HPLC)
≥94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text