Thermo Scientific Chemicals

Ethyl potassium oxalate, 97%, Thermo Scientific Chemicals

Catalog number: H35403.06
5 g, Each
Thermo Scientific Chemicals

Ethyl potassium oxalate, 97%, Thermo Scientific Chemicals

Catalog number: H35403.06
5 g, Each
Quantity
Catalog number: H35403.06
also known as H35403-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1906-57-6
IUPAC Name
potassium ethyl oxalate
Molecular Formula
C4H5KO4
InChI Key
RLPQQBNSTHRHEK-UHFFFAOYSA-M
SMILES
[K+].CCOC(=O)C([O-])=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

Ethyl potassium oxalate is used to prepare oxalic acid ethyl ester 2-oxo-3-phenyl-propyl ester by reacting with 1-bromo-3-phenyl-acetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl potassium oxalate is used to prepare oxalic acid ethyl ester 2-oxo-3-phenyl-propyl ester by reacting with 1-bromo-3-phenyl-acetone.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text