Thermo Scientific Chemicals

Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H35502.03
1 g, Each
Thermo Scientific Chemicals

Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H35502.03
1 g, Each
Quantity
Catalog number: H35502.03
also known as H35502-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
274-56-6
IUPAC Name
pyrazolo[1,5-a]pyridine
Molecular Formula
C7H6N2
InChI Key
DVUBDHRTVYLIPA-UHFFFAOYSA-N
SMILES
C1=NN2C=CC=CC2=C1
Form
Liquid
Appearance (Color)
Clear pale red to red to brown
Assay (GC)
≥96.0%
Refractive Index
1.6050-1.6100 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text