Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals
Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H35502.MD
also known as H35502-MD
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals
Catalog numberH35502.MD
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS274-56-6
IUPAC Namepyrazolo[1,5-a]pyridine
Molecular FormulaC7H6N2
InChI KeyDVUBDHRTVYLIPA-UHFFFAOYSA-N
SMILESC1=NN2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear pale red to red to brown
Assay (GC)≥96.0%
Refractive Index1.6050-1.6100 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only