tert-Butyl 4-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals
tert-Butyl 4-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl 4-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H35540.09
also known as H35540-09
Price (USD)/ Each
342.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
342.00
Add to cart
tert-Butyl 4-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals
Catalog numberH35540.09
Price (USD)/ Each
342.00
-
Add to cart
Chemical Identifiers
CAS108052-76-2
IUPAC Nametert-butyl 4-(bromomethyl)benzoate
Molecular FormulaC12H15BrO2
InChI KeyGSIBTIUXYYFCPU-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (GC)≥94.0%
Melting Point (clear melt)46.0-59.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only