3-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals
3-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H35558.03
also known as H35558-03
Price (USD)/ Each
194.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
194.00
Add to cart
3-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals
Catalog numberH35558.03
Price (USD)/ Each
194.00
-
Add to cart
Chemical Identifiers
CAS886371-28-4
IUPAC Name3-bromo-6-chloroimidazo[1,2-a]pyridine
Molecular FormulaC7H4BrClN2
InChI KeyLIBKDAUSMLPFBF-UHFFFAOYSA-N
SMILESClC1=CN2C(Br)=CN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)107.5-116.5?C
Appearance (Color)Pale brown
FormPowder
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only