Thermo Scientific Chemicals

Pyrrole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H35559.MD
250 mg, Each
Thermo Scientific Chemicals

Pyrrole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H35559.MD
250 mg, Each
Quantity
Catalog number: H35559.MD
also known as H35559-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7126-39-8
IUPAC Name
1H-pyrrole-3-carbaldehyde
Molecular Formula
C5H5NO
InChI Key
CHNYVNOFAWYUEG-UHFFFAOYSA-N
SMILES
O=CC1=CNC=C1
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Form
Powder
Free acid (titration)
≤1.5%
Melting Point (clear melt)
59.0-69.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text