6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H35567.MD
also known as H35567-MD
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals
Catalog numberH35567.MD
Price (USD)/ Each
161.00
-
Add to cart
Specifications
Chemical Name or Material6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate
CAS138642-97-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only