DAPI and Hoechst Nucleic Acid Stains
DAPI and Hoechst Nucleic Acid Stains
Invitrogen™

DAPI and Hoechst Nucleic Acid Stains

DAPI and Hoechst are classic popular nuclear counterstains for use in all fluorescent cell and tissue techniques. The blue fluorescence is vivid contrast to green, yellow, orange, red, far red and NIR fluorescent probes and labels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct TypeDye Type
H357010 mLNucleic Acid StainHoechst 33342, trihydrochloride trihydrate
622481 mLDAPIDAPI
D2149010 mgDAPIDAPI, FluoroPure grade
D357110 mgDAPIDAPI
D130610 mgDAPIDAPI
H356910 mLNucleic Acid StainHoechst 33258, pentahydrate (bis-benzimide)
H1398100 mgNucleic Acid StainHoechst 33258, pentahydrate (bis-benzimide)
H21491100 mgNucleic Acid StainHoechst 33258, pentahydrate (bis-benzimide), FluoroPure grade
622495 mLNucleic Acid StainHoechst 33342
H1399100 mgNucleic Acid StainHoechst 33342, trihydrochloride trihydrate
H21492100 mgNucleic Acid StainHoechst 33342, trihydrochloride trihydrate, FluoroPure grade
Catalog number H3570
Price (USD)
125.46
Special offer
Online exclusive
Ends: 27-Mar-2026
153.00
Save 27.54 (18%)
Each
Add to cart
Quantity:
10 mL
Product Type:
Nucleic Acid Stain
Dye Type:
Hoechst 33342, trihydrochloride trihydrate
Price (USD)
125.46
Special offer
Online exclusive
Ends: 27-Mar-2026
153.00
Save 27.54 (18%)
Each
Add to cart

DAPI is a fluorescent dye commonly used in molecular biology and cell biology research. DAPI is known for its ability to bind to DNA, specifically to the minor groove of double-stranded DNA. This dye emits blue fluorescence when excited by ultraviolet light, allowing researchers to visualize and stain DNA in various biological samples.

Hoechst dyes are DNA-specific fluorescent stains that bind to the minor groove of double-stranded DNA. They emit blue fluorescence when excited by ultraviolet light. These dyes are widely used for staining DNA in various biological samples.

The FluoroPure™-grade of Hoechst and DAPI powders are manufactured using stringent purification processes to minimize impurities that could interfere with fluorescence imaging. This ensures that the DAPI and Hoechst provides reliable and consistent results with minimal background noise. The >98% purity of FluoroPure grade reagents make them particularly suitable for demanding applications such as super-resolution microscopy and live-cell imaging, where precise and accurate visualization of DNA is essential.

DAPI (4',6-Diamidino-2-Phenylindole):

DAPI 4',6-diamidino-2-phenylindole is a blue fluorescent probe that fluoresces brightly upon selectively binding to the minor groove of double stranded DNA, where its fluorescence is approximately 20-fold greater than in the non-bound state. Its selectivity for DNA and high cell permeability allows efficient staining of nuclei with little background from the cytoplasm and efficient contrasting in IHC staining and spatial transcriptomics and proteomics tissue imaging. DAPI is a classic nuclear counterstain for immunofluorescence microscopy, as well as an important component of high-content screening methods requiring cell-based quantitation of DNA content and segmentation (ex/em 341/452 nm).

DAPI as a dilactate powder is more water soluble than the dihydrochloride salt, and therefore a better choice for preparing stock solutions in water.

To make a 5 mg/mL DAPI stock solution (14.3 mM for the dihydrochloride or 10.9 mM for the dilactate), dissolve the contents of one vial (10 mg) in 2 mL of deionized water (dH2O) or dimethylformamide (DMF). The less water-soluble DAPI dihydrochloride may take some time to completely dissolve in water and sonication may be necessary.

Note: Neither of these DAPI derivatives is very soluble in PBS for Stock Solution.

Dilute the DAPI Stock Solution 1:5000 (or 1:1000 for the 1 mg/mL DAPI 62248) in ultrapure water or PBS (1 μg/mL DAPI). Filter the working solution to remove dye aggregates that can result in punctate signal.

Hoechst 33342, Trihydrochloride, Trihydrate and Hoechst 33258, Pentahydrate (bis-Benzimide):

Our Hoechst dyes are renowned fluorophores widely used for DNA staining in both living and fixed cells. With their exceptional affinity and specificity towards DNA, these dyes serve as excellent targeting agents that can be conjugated to various molecules, allowing them to be tethered to DNA.

The Hoechst dyes typically include Hoechst 33258, Hoechst 33342, and Hoechst 34580. Developed by the German company Hoechst AG in the early 1970s, these bisbenzimide dyes have been widely used for DNA staining ever since.

Excited by UV light, Hoechst dyes emit a broad spectrum of blue light. Upon binding to DNA, their fluorescence increases approximately 30-fold, ensuring a strong signal-to-noise ratio. This fluorescence enhancement is the result of suppressed rotational relaxation and hydration reduction upon DNA binding (ex/em 360/460 nm).

Hoechst dyes are non-intercalating, binding specifically to the minor groove of DNA at A–T-rich regions. This unique binding mechanism allows for precise and reliable DNA staining.

Whether you're working with living or fixed cells, our Hoechst dyes are compatible with immunohistochemistry applications. Additionally, the binding of Hoechst 33342 to DNA induces minimal cytotoxicity, ensuring the viability of your cells.

While all three Hoechst dyes have similar applications, they do possess slight differences in their properties. Hoechst 33342, for example, is significantly more cell-permeable due to the addition of a lipophilic ethyl group, making it the preferred choice for living cell staining. On the other hand, Hoechst 34580, with its dimethylamine group instead of the phenol, exhibits a shifted emission maximum at 490 nm, compared to the 461 nm of Hoechst 33258 and Hoechst 33342.

Hoechst 33258, Pentahydrate is also known as bis-Benzimide.

Application of DAPI and Hoechst Fluorescent Dyes:

  • DNA Staining—commonly used for DNA staining in fluorescence microscopy and flow cytometry. It binds to the minor groove of double-stranded DNA and emits blue fluorescence when excited by ultraviolet (UV) light. This allows researchers to visualize and quantify DNA in cells, tissues, and nuclei.
  • Cell Cycle Analysis—widely used to analyze the cell cycle. By staining cells with this dye and using flow cytometry or fluorescence microscopy, researchers can identify and quantify different phases of the cell cycle, such as G1, S, and G2/M phases. This analysis provides insights into cell proliferation, cell cycle progression, and cell cycle abnormalities.
  • Nuclear Morphology and Chromatin Organization—study nuclear morphology and chromatin organization. By visualizing the DNA within the nucleus, researchers can investigate changes in nuclear size, shape, and chromatin condensation, which are associated with various cellular processes and conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue
Concentration10 mg/mL
Detection MethodFluorescence
Dye TypeHoechst 33342, trihydrochloride trihydrate
Emission461 nm
Excitation Wavelength Range350 nm
For Use With (Application)Fluorescent labeling
For Use With (Equipment)Fluorescence Microscope
FormLiquid
Product LineHoechst
Quantity10 mL
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeNucleic Acid Stain
SubCellular LocalizationNucleic Acids, Nucleus
Unit SizeEach
Contents & Storage
Store in refrigerator at 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

Is DAPI a good live-cell nuclear label?

DAPI is considered a semi-permeant/impermeant nucleic acid stain. Staining of nucleic is dependent upon the cell line in its performance. Some cell lines will label with DAPI, others not at all, and others label inconsistenly. Instead, we recommend using either Hoechst 33342 or Hoechst 33258, which have the same wavelength and binding mode as DAPI (at the A-T minor groove) but are readily cell-permeant.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need a total cell stain, similar to HCS CellMask Blue stain, to label cytoplasm and nuclei in live cells. What do you recommend?

You can use a combination of two dyes with overlapping blue emission. For cytoplasm, you can label the cell with CellTracker Blue CMAC. This can be combined with Hoechst 33342 for nuclei. Both dyes are imaged using a standard DAPI filter set.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to label the nuclei of live cells and track them over time. Can I use DAPI for this?

We do not recommend doing this. DAPI is considered to be a semi-permeant/impermeant nucleic acid stain. DAPI staining of live cells may be inconsistent. It is best used as a counterstain for fixed samples. Other cell permeable nucleic acid stains, such as Hoechst or the SYTO dyes may affect cellular function.

For mammalian cells, we recommend using the CellLight Nucleus transduction reagents, available in CFP, GFP and RFP. With these reagents, the cells are transduced overnight in a single labeling step and the next day the nuclei will fluoresce. The label may be retained for 3-5 days and should not affect cell function. Cytoplasmic cell tracking dyes such as the CellTracker dyes may also be used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I have a mix of live bacteria and platelet cells, and I need to be able to separate out the bacteria. Do you have a suggestion?

Platelet cells don't have DNA, while bacteria do. Therefore, a cell-permeant, DNA-selective dye would preferentially stain the bacteria with limited staining of the platelets. We recommend using Hoechst 33342 dye.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

DAPI and Hoechst dyes are quite similar to each other. Why would I choose one over the other?

DAPI is a very common blue-fluorescent dye for nuclear counterstaining and gives very bright labeling on nuclei in fixed and permeabilized cells and tissues. However, it is considered to be a semi-permeant to impermeant stain and provides inconsistent staining of live cells. Hoechst 33342 dye is cell-permeant and stains with the same binding mechanism and fluorescent color; it is preferred for live-cell imaging and is just as good as DAPI for fixed cell labeling.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (639)

Citations & References
Abstract
Hematopoietic potential of stem cells isolated from murine skeletal muscle.
Authors:Jackson KA,Mi T,Goodell MA
Journal:Proceedings of the National Academy of Sciences of the United States of America
PubMed ID:10588731
We have discovered that cells derived from the skeletal muscle of adult mice contain a remarkable capacity for hematopoietic differentiation. Cells prepared from muscle by enzymatic digestion and 5-day in vitro culture were harvested, and 18 × 10(3) cells were introduced into each of six lethally irradiated recipients together with ... More
P-glycoprotein-mediated Hoechst 33342 transport out of the lipid bilayer.
Authors:Shapiro AB, Corder AB, Ling V
Journal:Eur J Biochem
PubMed ID:9431998
High-level expression of P-glycoprotein, a 170-kDa mammalian plasma membrane ATPase, is the cause of an important and widespread form of cancer multidrug resistance. P-glycoprotein reduces cellular accumulation of an enormous variety of lipophilic compounds. The basis for this broad substrate specificity is not well understood. We explored this issue by ... More
Assembly of the nuclear pore: biochemically distinct steps revealed with NEM, GTP gamma S, and BAPTA.
Authors:Macaulay C, Forbes DJ
Journal:J Cell Biol
PubMed ID:8567730
A key event in nuclear formation is the assembly of functional nuclear pores. We have used a nuclear reconstitution system derived from Xenopus eggs to examine the process of nuclear pore assembly in vitro. With this system, we have identified three reagents which interfere with nuclear pore assembly, NEM, GTP ... More
High-speed photodamage cell selection using bromodeoxyuridine/Hoechst 33342 photosensitized cell killing.
Authors:Herweijer H, Stokdijk W, Visser JW
Journal:Cytometry
PubMed ID:2452055
One of the major drawbacks of droplet sorting in a flow cytometer is the relatively low sorting speed. Thus, we have developed an alternative, faster sorting technique: photodamage cell sorting. In a photodamage cell sorter all unwanted cells, as detected with the first, measuring laser, are killed with the second, ... More
Cyclin G2 associates with protein phosphatase 2A catalytic and regulatory B' subunits in active complexes and induces nuclear aberrations and a G1/S phase cell cycle arrest.
Authors:Bennin David A; Don Aruni S Arachchige; Brake Tiffany; McKenzie Jennifer L; Rosenbaum Heidi; Ortiz Linette; DePaoli-Roach Anna A; Horne Mary C;
Journal:J Biol Chem
PubMed ID:11956189
Cyclin G2, together with cyclin G1 and cyclin I, defines a novel cyclin family expressed in terminally differentiated tissues including brain and muscle. Cyclin G2 expression is up-regulated as cells undergo cell cycle arrest or apoptosis in response to inhibitory stimuli independent of p53 (Horne, M., Donaldson, K., Goolsby, G., ... More