6-Fluoroquinoline, 97%, Thermo Scientific Chemicals
6-Fluoroquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Fluoroquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H35900.MD
also known as H35900-MD
Price (USD)/ Each
63.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
63.00
Add to cart
6-Fluoroquinoline, 97%, Thermo Scientific Chemicals
Catalog numberH35900.MD
Price (USD)/ Each
63.00
-
Add to cart
Chemical Identifiers
CAS396-30-5
IUPAC Name6-fluoroquinoline
Molecular FormulaC9H6FN
InChI KeyRMDCSDVIVXJELQ-UHFFFAOYSA-N
SMILESFC1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5905-1.5955 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only