Thermo Scientific Chemicals

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H35994.03
1 g, Each
Thermo Scientific Chemicals

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H35994.03
1 g, Each
Quantity
Catalog number: H35994.03
also known as H35994-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
342613-63-2
IUPAC Name
methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Molecular Formula
C9H7BrN2O2
InChI Key
VFFFBWRUZMAHFS-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC2=NC=C(Br)N2C=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥94.0%
Appearance (Color)
White
Melting Point (clear melt)
156.5-165.5?C

Description

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text