Thermo Scientific Chemicals

Platinum, 10% on activated carbon paste, Type 128M, Thermo Scientific Chemicals

Catalog number: H36044.14
25 g, Each
Thermo Scientific Chemicals

Platinum, 10% on activated carbon paste, Type 128M, Thermo Scientific Chemicals

Catalog number: H36044.14
25 g, Each
Quantity
Catalog number: H36044.14
also known as H36044-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-06-4
IUPAC Name
platinum
Molecular Formula
Pt
InChI Key
BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES
[Pt]
Loss on Drying
45-65%
Appearance (Color)
Black
Elemental Analysis
Platinum, Pt: 9 -11%
Form
Solid

Description

Platinum as a catalyst is used for hydrogenation of alkenes, alkynes, nitro groups. It is also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum as a catalyst is used for hydrogenation of alkenes, alkynes, nitro groups. It is also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Insoluble in water, mineral acids, and organic acids. Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, organic materials and flammable substances.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text