Thermo Scientific Chemicals

1,3,5-Benzenetriboronic acid tris(pinacol) ester, 97%, Thermo Scientific Chemicals

Catalog number: H36045.06
5 g, Each
Thermo Scientific Chemicals

1,3,5-Benzenetriboronic acid tris(pinacol) ester, 97%, Thermo Scientific Chemicals

Catalog number: H36045.06
5 g, Each
Quantity
Catalog number: H36045.06
also known as H36045-06
Price (USD)
Quantity
-
Have Questions?
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Appearance (Color)
White to cream or pale brown

Description

1. Fabricated a 2D COF which when deposited onto a Gold electrode increases the hydrogen evolution reaction(HER) activity by up to three times compared with bare Gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text