Chemical Identifiers
CAS136040-19-2
IUPAC Nametetrakis(2,3,4,5,6-pentafluorophenyl)boranuide; triphenylmethylium
Molecular FormulaC43H15BF20
InChI KeyTZOSNOQHGGONMD-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
View more