Thermo Scientific Chemicals

1-Octanol, natural, 98%, Thermo Scientific Chemicals

Catalog number: H36188.AP
500 mL, Each
Thermo Scientific Chemicals

1-Octanol, natural, 98%, Thermo Scientific Chemicals

Catalog number: H36188.AP
500 mL, Each
Quantity
Catalog number: H36188.AP
also known as H36188-AP
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
111-87-5
IUPAC Name
octan-1-ol
Molecular Formula
C8H18O
InChI Key
KBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILES
CCCCCCCCO
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4270-1.4310 @ 20?C

Description

1-Octanol is a synthetic reagent used to produce esters. It inhibits T-type calcium channels and is used to determine the partition coefficient between compounds and the cytosol. It is used to evaluate the lipophilicity of pharmaceutical products. It is also used as a solvent for protective coatings, waxes, and oils, and as a raw material for plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is a synthetic reagent used to produce esters. It inhibits T-type calcium channels and is used to determine the partition coefficient between compounds and the cytosol. It is used to evaluate the lipophilicity of pharmaceutical products. It is also used as a solvent for protective coatings, waxes, and oils, and as a raw material for plasticizers.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 2
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H319 - Causes serious eye irritation

    H412 - Harmful to aquatic life with long lasting effects

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear eye protection/ face protection

    P337 + P313 - If eye irritation persists: Get medical advice/attention