Thermo Scientific Chemicals

Rhodium, 5% on alumina powder, C301099-5, Thermo Scientific Chemicals

Catalog number: H36201.06
5 g, Each
Thermo Scientific Chemicals

Rhodium, 5% on alumina powder, C301099-5, Thermo Scientific Chemicals

Catalog number: H36201.06
5 g, Each
Quantity
Catalog number: H36201.06
also known as H36201-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-16-6
IUPAC Name
rhodium
Molecular Formula
Rh
InChI Key
MHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES
[Rh]
Appearance (Color)
Grey
Moisture holding capacity
≤5.0 Wt.%
Form
Powder
Elemental Analysis
(Rhodium, Rh) : 4.800-5.100 Wt.%

Description

Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with organic materials and flammable substances.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text