Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with organic materials and flammable substances.
RUO – Research Use Only
Joseph J. Mrowca.; Thomas J. Katz. Catalysis of a Cycloaddition Reaction by Rhodium on Carbon. J. Am. Chem. Soc. 1966, 88,(17), 4012-4015.
Morris Freifelder.; Ralph M. Robinson.; George R. Stone. Hydrogenation of Substituted Pyridines with Rhodium on Carbon Catalyst. J. Org. Chem. 1962, 27,(1), 284-286.