Thermo Scientific Chemicals

Lithium hydroxide monohydrate, battery grade, Thermo Scientific Chemicals

Catalog number: H36379.30
250 g, Each
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, battery grade, Thermo Scientific Chemicals

Catalog number: H36379.30
250 g, Each
Quantity
Catalog number: H36379.30
also known as H36379-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-66-3
IUPAC Name
lithium(1+) hydrate hydroxide
Molecular Formula
H3LiO2
InChI Key
GLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES
[Li+].O.[OH-]
Chloride content
≤500ppm (≤0.05%)
Impurity content
CO2: ≤3500ppm (≤0.35%)
Impurity content
Iron, Fe: ≤10ppm (≤0.001%)
Impurity content
Sulphate, SO4: ≤500ppm (≤0.005%)
Insoluble material
≤0.1% (in water)

Description

Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

Solubility
Soluble in water (216 mg/ml at 20°C), ethanol (freely), and alcohol (slightly).

Notes
Air sensitive and hygroscopic. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text