4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone, Thermo Scientific Chemicals
4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Catalog number H36621.AC
also known as H36621-AC
Price (USD)/ Each
50.60
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
50.60
Add to cart
4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone, Thermo Scientific Chemicals
Catalog numberH36621.AC
Price (USD)/ Each
50.60
-
Add to cart
Chemical Identifiers
CAS100-43-6
IUPAC Name4-ethenylpyridine
Molecular FormulaC7H7N
InChI KeyKFDVPJUYSDEJTH-UHFFFAOYSA-N
SMILESC=CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to red/brown
FormLiquid
Assay (GC)≥95.0%
CommentMay darken on storage
Refractive Index1.5480-1.5540 @20?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only