Thermo Scientific Chemicals

4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: H36621.AE
100 mL, Each
Thermo Scientific Chemicals

4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: H36621.AE
100 mL, Each
Quantity
Catalog number: H36621.AE
also known as H36621-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-43-6
IUPAC Name
4-ethenylpyridine
Molecular Formula
C7H7N
InChI Key
KFDVPJUYSDEJTH-UHFFFAOYSA-N
SMILES
C=CC1=CC=NC=C1
Appearance (Color)
Clear colorless to yellow to red/brown
Form
Liquid
Assay (GC)
≥95.0%
Comment
May darken on storage
Refractive Index
1.5480-1.5540 @20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text