Thermo Scientific Chemicals

1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: H36633.03
1 g, Each
Thermo Scientific Chemicals

1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: H36633.03
1 g, Each
Quantity
Catalog number: H36633.03
also known as H36633-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
286014-38-8
IUPAC Name
1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
Molecular Formula
C15H25BF4N2
InChI Key
CQHXJIHJFMBBQA-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1
Elemental Analysis
Carbon C: 54.02 - 58.52% (Theory 56.27%)
Proton NMR
Conforms to structure
Appearance (Color)
White to pale cream
Form
Powder
Assay (unspecified)
> 96.0% by Carbon EA

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text