1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals
1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H36633.06
also known as H36633-06
Price (USD)/ Each
363.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
363.00
Add to cart
1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Catalog numberH36633.06
Price (USD)/ Each
363.00
-
Add to cart
Chemical Identifiers
CAS286014-38-8
IUPAC Name1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
Molecular FormulaC15H25BF4N2
InChI KeyCQHXJIHJFMBBQA-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisCarbon C: 54.02 - 58.52% (Theory 56.27%)
Proton NMRConforms to structure
Appearance (Color)White to pale cream
FormPowder
Assay (unspecified)> 96.0% by Carbon EA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only