5-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals
5-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H36784.MD
also known as H36784-MD
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
5-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberH36784.MD
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS31555-59-6
IUPAC Name5-methylthiophene-2-carbonyl chloride
Molecular FormulaC6H5ClOS
InChI KeyDJCGIWVUKCNCAT-UHFFFAOYSA-N
SMILESCC1=CC=C(S1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (Titration ex Chloride)> 97.5%
Assay (GC)> 97.5%
Refractive Index1.5835 - 1.5875 @20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only