Thermo Scientific Chemicals

Ramipril, 98%, Thermo Scientific Chemicals

Catalog number: H36926.MD
250 mg, Each
Thermo Scientific Chemicals

Ramipril, 98%, Thermo Scientific Chemicals

Catalog number: H36926.MD
250 mg, Each
Size
Catalog number: H36926.MD
also known as H36926-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87333-19-5
IUPAC Name
(2S,3aS,6aS)-1-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-octahydrocyclopenta[b]pyrrole-2-carboxylic acid
Molecular Formula
C23H32N2O5
InChI Key
HDACQVRGBOVJII-JBDAPHQKSA-N
SMILES
CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1[C@H]2CCC[C@H]2C[C@H]1C(O)=O
Appearance (Color)
White to pale cream
Optical Rotation
+15.0 ± 2.0? (c=1, methanol)
Assay (HPLC)
≥97.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text