Thermo Scientific Chemicals

Fosinopril (sodium), 98%, Thermo Scientific Chemicals

Catalog number: H37120.MD
250 mg, Each
Thermo Scientific Chemicals

Fosinopril (sodium), 98%, Thermo Scientific Chemicals

Catalog number: H37120.MD
250 mg, Each
Quantity
Catalog number: H37120.MD
also known as H37120-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88889-14-9
IUPAC Name
sodium (2S,4S)-4-cyclohexyl-1-(2-{[2-methyl-1-(propanoyloxy)propoxy](4-phenylbutyl)phosphoryl}acetyl)pyrrolidine-2-carboxylate
Molecular Formula
C30H45NNaO7P
InChI Key
TVTJZMHAIQQZTL-HREVRLCXSA-M
SMILES
[Na+].CCC(=O)OC(OP(=O)(CCCCC1=CC=CC=C1)CC(=O)N1C[C@@H](C[C@H]1C([O-])=O)C1CCCCC1)C(C)C
Assay from Supplier's CofA
≥97.5% (HPLC, US-sourced material)
Mass Spec
Conforms (US-sourced material)
Appearance (Color)
White
Form
Powder
Assay (HPLC)
≥97.5% (UK-sourced material)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text